2-Fluor-4-(trifluormethyl)benzonitril

Lieferant: FLUOROCHEM

Synonyme: 2-Fluor-4-(trifluormethyl)benzoesäurenitril, 4-Cyan-3-fluorbenzotrifluorid

002015-1G 002015-5G
FLUO002015-1GEA 31.1 EUR
FLUO002015-1G FLUO002015-5G
2-Fluor-4-(trifluormethyl)benzonitril
2-Fluor-4-(trifluormethyl)benzonitril
Formel: C₈H₃F₄N
Molecular Weight: 189,11 g/mol
Siedepunkt: 178 °C (1013 hPa)
Dichte: 1,35 g/cm³ (20 °C)
MDL: MFCD00061279
CAS-Nummer: 146070-34-0
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR