2-Fluor-5-(trifluormethyl)benzonitril
Lieferant: FLUOROCHEM
Synonyme:
2-Fluor-5-(trifluormethyl)benzoesäurenitril, 3-Cyan-4-fluorbenzotrifluorid
002030-1G
002030-5G
002030-25G
FLUO002030-1GEA
31.1
EUR
FLUO002030-1G
FLUO002030-5G
FLUO002030-25G
2-Fluor-5-(trifluormethyl)benzonitril
2-Fluor-5-(trifluormethyl)benzonitril
Formel:
C₈H₃F₄N Molecular Weight: 189.11 g/mol Siedepunkt: 99 |
MDL:
MFCD00061280 CAS-Nummer: 4088-84-0 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...