4-Fluor-3-(trifluormethyl)benzonitril
Lieferant: CHEMPUR
Synonyme:
5-Cyano-2-fluorobenzotrifluoride
CFF1155.
FL2223.1
002223-5G
002223-25G
CHMPCFF1155.EA
0
EUR
CHMPCFF1155.
CHMPFL2223.1
FLUO002223-5G
FLUO002223-25G
4-Fluor-3-(trifluormethyl)benzonitril
4-Fluor-3-(trifluormethyl)benzonitril
Formel:
C₈H₃F₄N Molecular Weight: 189.11 g/mol Siedepunkt: 193…195 °C (1013 hPa) Schmelzpunkt: 67…68 °C |
MDL:
MFCD00061284 CAS-Nummer: 67515-59-7 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...