1-(4-Nitrophenyl)glycerin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(4-Nitrophenyl)-1,2,3-propantriol, PNPG

N1143-1G N1143-5G
SIALN1143-1GEA 190 EUR
SIALN1143-1G SIALN1143-5G
1-(4-Nitrophenyl)glycerin, Sigma-Aldrich®
1-(4-Nitrophenyl)glycerin
Formel: C₉H₁₁NO₅
Molecular Weight: 213,19 g/mol
Schmelzpunkt: 94…97 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00017037
CAS-Nummer: 2207-68-3
EINECS: 218-624-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR