D(−)-Salicin 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
α-Hydroxy-o-tolyl-β-D-glucopyranosid, (2R3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxan-3,4,5-triol, Weidenbitter, Salicinum, Saligenin-β-D-glucopyranosid, 2-(Hydroxymethyl)phenyl-beta-D-glucopyranosid, Salicosid
132590250
132591000
132590050
ACRO132590250EA
212
EUR
ACRO132590250
ACRO132591000
ACRO132590050
D(−)-Salicin 99+%
D(−)-Salicin
Formel:
C₁₃H₁₈O₇ Molecular Weight: 286.28 g/mol Schmelzpunkt: 197…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006590 CAS-Nummer: 138-52-3 EINECS: 205-331-6 Merck Index: 12,08476 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % |
HPLC | ≥99.0 % |
Specific optical rotation | -59.5° to -63.5° (20°C, 589 nm) (c=5, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...