D(−)-Salicin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: α-Hydroxy-o-tolyl-β-D-glucopyranosid, (2R3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxan-3,4,5-triol, Weidenbitter, Salicinum, Saligenin-β-D-glucopyranosid, 2-(Hydroxymethyl)phenyl-beta-D-glucopyranosid, Salicosid

M-Clarity™ Qualitätsstufe = MQ200

S0625-100G S0625-25G S0625-5G
SIALS0625-100GEA 1060 EUR
SIALS0625-100G SIALS0625-25G SIALS0625-5G
D(−)-Salicin, Sigma-Aldrich®
D(−)-Salicin
Formel: C₁₃H₁₈O₇
Molecular Weight: 286,28 g/mol
Schmelzpunkt: 199…201 °C
Dichte: 1,42 g/cm³ (20 °C)
MDL: MFCD00006590
CAS-Nummer: 138-52-3
EINECS: 205-331-6
Merck Index: 12,08476

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR