Alizarin 97%, rein
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,2-Dihydroxy-9,10-anthrachinon, 1,2-Dihydroxyanthrachinon
153690250
153691000
153695000
153690025
ACRO153690250EA
42.9
EUR
ACRO153690250
ACRO153691000
ACRO153695000
ACRO153690025
Alizarin 97%, rein
Alizarin
Formel:
C₁₄H₈O₄ Molecular Weight: 240,22 g/mol Siedepunkt: 430 °C (1013 hPa) Schmelzpunkt: 287…289 °C Dichte: 1,54 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001201 CAS-Nummer: 72-48-0 EINECS: 200-782-5 Merck Index: 13,00246 |
Spezifikation Testergebnisse
Appearance | Orange to orange-brown Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % |
Dye content | ≥96.0 % |
Residue after evaporation | ≤0.002 % |
Acidity | ≤0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...