4-Indolcarbaldehyd

Lieferant: Cayman Chemical

Synonyme: Indol-4-carboxaldehyd

10007086-1 10007086-10 10007086-5 10007086-500
CAYM10007086-1EA 189 EUR
CAYM10007086-1 CAYM10007086-10 CAYM10007086-5 CAYM10007086-500
4-Indolcarbaldehyd
4-Indolcarbaldehyd
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
MDL: MFCD01632221
CAS-Nummer: 1074-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR