3-Amino-1H-1,2,4-triazol 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Amino-1,2,4-triazol, Amitrol, 3-Amino-1H-1,2,4-triazol
264571000
264570100
ACRO264571000EA
138
EUR
ACRO264571000
ACRO264570100
3-Amino-1H-1,2,4-triazol 95%
3-Amino-1H-1,2,4-triazol
Formel:
C₂H₄N₄ Molecular Weight: 84.08 g/mol Siedepunkt: 347 °C (1013 hPa) Schmelzpunkt: 149…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053362 CAS-Nummer: 61-82-5 EINECS: 200-521-5 UN: 3077 ADR: 9 III Merck Index: 13,00489 |
Spezifikation Testergebnisse
Appearance | White to off-white to yellow Powder or flakes |
Infrared spectrum | Conforms |
Melting point | 147 to 156 °C |
Titration with HClO4 | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...