1,2,3-Triazol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,2,3-1H-Triazol

A16999.03 A16999.06 A16999.14
A16999.03EA 58 EUR
A16999.03 A16999.06 A16999.14
1,2,3-Triazol ≥98%
1,2,3-Triazole

Purity: 98% CAS No.: 288-36-8 Molecular Formula: C2H3N3 Formula Weight: 69.07 Flash Point: 107°(224°F) Refractive Index: 1.4980 Melting Point: 23-25° Boiling Point: 208° Beilstein Reference: 104766 MDL No.: MFCD00014490

Formel: C₂H₃N₃
Molecular Weight: 69,07 g/mol
Siedepunkt: 208 °C
Schmelzpunkt: 23…25 °C
Dichte: 1,192
Flammpunkt: 107 °C (224 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014490
CAS-Nummer: 288-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR