4-Amino-4H-1,2,4-triazol 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Amino-1,2,4-triazol, 1,2,4-Triazol-4-ylamin

A17716.06 A17716.14 A17716.22
A17716.06EA 27.3 EUR
A17716.06 A17716.14 A17716.22
4-Amino-4H-1,2,4-triazol 99%
4-Amino-4H-1,2,4-triazol

Purity: 99% CAS No.: 584-13-4 Molecular Formula: C2H4N4 Formula Weight: 84.08 Melting Point: 83-88° Beilstein Reference: 107563

Formel: C₂H₄N₄
Molecular Weight: 84.08 g/mol
Schmelzpunkt: 83…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003099
CAS-Nummer: 584-13-4
EINECS: 209-533-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR