8-Chlorchinaldin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 8-Chloro-2-methylquinoline

B20504.03 B20504.06
B20504.03EA 28.8 EUR
B20504.03 B20504.06
8-Chlorchinaldin ≥98%
8-Chlorchinaldin

Purity: 98% CAS No.: 3033-82-7 Molecular Formula: C10H8ClN Formula Weight: 177.63 Melting Point: 65-68° Beilstein Reference: 119193

Formel: C₁₀H₈ClN
Molecular Weight: 177.63 g/mol
Schmelzpunkt: 65…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051764
CAS-Nummer: 3033-82-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR