3,5-Diamino-1H-1,2,4-triazol ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Guanazol

B22775.06 B22775.14 B22775.22
B22775.06EA 31.2 EUR
B22775.06 B22775.14 B22775.22
3,5-Diamino-1H-1,2,4-triazol ≥98%
3,5-Diamino-1H-1,2,4-triazol

Purity: 98+% CAS No.: 1455-77-2 Molecular Formula: C2H5N5 Formula Weight: 99.10 Melting Point: ca 205° dec. Beilstein Reference: 112467

Formel: C₂H₅N₅
Molecular Weight: 99.1 g/mol
Siedepunkt: 425
Schmelzpunkt: ∼ 205 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005233
CAS-Nummer: 1455-77-2
EINECS: 215-937-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR