4'-Methyl-2,2,2-trifluoracetophenon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,2,2-Trifluor-1-(4-methylphenyl)-1-ethanon
L12424.03
L12424.06
L12424.03EA
64.4
EUR
L12424.03
L12424.06
4'-Methyl-2,2,2-trifluoracetophenon ≥97%
4'-Methyl-2,2,2-trifluoracetophenon
Purity: 97%
CAS No.: 394-59-2
Molecular Formula: C9H7F3O
Formula Weight: 188.15
Refractive Index: 1.4700
Boiling Point: 70-72°/15mm
MDL No.: MFCD00052338
Formel:
C₉H₇F₃O Molecular Weight: 188,15 g/mol Siedepunkt: 70…72 °C (15 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052338 CAS-Nummer: 394-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...