3-Nitro-1H-1,2,4-triazol

Lieferant: Thermo Scientific

Synonyme: 3-Nitro-1,2,4-1H-triazol

JFD01290.250 JFD01290.1
MAYBJFD01290.250EA 0 EUR
MAYBJFD01290.250 MAYBJFD01290.1
3-Nitro-1H-1,2,4-triazol
3-Nitro-1H-1,2,4-triazol
Formel: C₂H₂N₄O₂
Molecular Weight: 114.06 g/mol
Schmelzpunkt: 210…215 °C
Dichte: 1.65 g/cm³ (20 °C)
MDL: MFCD00009749
CAS-Nummer: 24807-55-4
EINECS: 246-468-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR