6-Bromchinaldin

Lieferant: FLUOROCHEM

002521-1G 002521-5G 002521-25G
FLUO002521-1GEA 0 EUR
FLUO002521-1G FLUO002521-5G FLUO002521-25G
6-Bromchinaldin
6-Bromchinaldin
Formel: C₁₀H₈BrN
Molecular Weight: 222,08 g/mol
MDL: MFCD00079724
CAS-Nummer: 877-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR