7-Chlorchinaldin

Lieferant: FLUOROCHEM

Synonyme: 7-Chlor-2-methylchinolin

002536-5G 002536-25G
FLUO002536-5GEA 0 EUR
FLUO002536-5G FLUO002536-25G
7-Chlorchinaldin
7-Chlorchinaldin
Formel: C₁₀H₈ClN
Molecular Weight: 177.63 g/mol
MDL: MFCD00075436
CAS-Nummer: 4965-33-7
EINECS: 440-600-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR