4'-Methyl-2,2,2-trifluoracetophenon

Lieferant: CHEMPUR

Synonyme: 2,2,2-Trifluor-1-(4-methylphenyl)-1-ethanon

FL7475.POR 007475-1G 007475-5G
CHMPFL7475.POREA 0 EUR
CHMPFL7475.POR FLUO007475-1G FLUO007475-5G
4'-Methyl-2,2,2-trifluoracetophenon
4'-Methyl-2,2,2-trifluoracetophenon
Formel: C₉H₇F₃O
Molecular Weight: 188.15 g/mol
MDL: MFCD00052338
CAS-Nummer: 394-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR