4-Amino-4H-1,2,4-triazol

Lieferant: FLUOROCHEM

Synonyme: 4-Amino-1,2,4-triazol, 1,2,4-Triazol-4-ylamin

013753-25G 013753-100G
FLUO013753-25GEA 42.9 EUR
FLUO013753-25G FLUO013753-100G
4-Amino-4H-1,2,4-triazol
4-Amino-4H-1,2,4-triazol
Formel: C₂H₄N₄
Molecular Weight: 84.08 g/mol
MDL: MFCD00003099
CAS-Nummer: 584-13-4
EINECS: 209-533-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR