6-Bromchinaldin, Sigma-Aldrich®

Lieferant: Merck

649279-25G 649279-5G
SIAL649279-25GEA 123 EUR
SIAL649279-25G SIAL649279-5G
6-Bromchinaldin, Sigma-Aldrich®
6-Bromchinaldin
Formel: C₁₀H₈BrN
Molecular Weight: 222,08 g/mol
MDL: MFCD00079724
CAS-Nummer: 877-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR