3-Iod-4-nitrotoluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

659320-25G 659320-5G
SIAL659320-25GEA 317 EUR
SIAL659320-25G SIAL659320-5G
3-Iod-4-nitrotoluol, Sigma-Aldrich®
3-Iod-4-nitrotoluol
Formel: C₇H₆INO₂
Molecular Weight: 263,03 g/mol
Schmelzpunkt: 95…97 °C
MDL: MFCD01320681
CAS-Nummer: 52488-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR