4-Amino-4H-1,2,4-triazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Amino-1,2,4-triazol, 1,2,4-Triazol-4-ylamin
M-Clarity™ Qualitätsstufe = MQ200
A81803-100G
A81803-25G
A81803-5G
SIALA81803-100GEA
110
EUR
SIALA81803-100G
SIALA81803-25G
SIALA81803-5G
4-Amino-4H-1,2,4-triazol, Sigma-Aldrich®
4-Amino-4H-1,2,4-triazol
Formel:
C₂H₄N₄ Molecular Weight: 84.08 g/mol |
MDL:
MFCD00003099 CAS-Nummer: 584-13-4 EINECS: 209-533-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...