Triacetoxyethylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Ethylsilantriyltriacetat, Triacetoxy(ethyl)silan

SILS08800.25 SILS08800.100
SILNSILS08800.25EA 0 EUR
SILNSILS08800.25 SILNSILS08800.100
Triacetoxyethylsilan
Triacetoxyethylsilan
Formel: C₈H₁₄O₆Si
Molecular Weight: 234.28 g/mol
MDL: MFCD00053703
CAS-Nummer: 17689-77-9
EINECS: 241-677-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR