Triacetoxymethylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Triacetoxy(methyl)silan, Methylsilantriyltriacetat

SILS11725.10 SILS11725.25 SILS11725.100
SILNSILS11725.10EA 0 EUR
SILNSILS11725.10 SILNSILS11725.25 SILNSILS11725.100
Triacetoxymethylsilan
Triacetoxymethylsilan
Formel: C₇H₁₂O₆Si
Molecular Weight: 220.25 g/mol
MDL: MFCD00008694
CAS-Nummer: 4253-34-3
EINECS: 224-221-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR