Triacetoxyvinylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Triacetoxy(vinyl)silan, Vinylsilantriyltriacetat

SILS21925.25 SILS21925.50
SILNSILS21925.25EA 0 EUR
SILNSILS21925.25 SILNSILS21925.50
Triacetoxyvinylsilan
Triacetoxyvinylsilan
Formel: C₈H₁₂O₆Si
Molecular Weight: 232.27 g/mol
MDL: MFCD00014975
CAS-Nummer: 4130-08-9
EINECS: 223-943-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR