Test tube racks, Azlon®
Lieferant: SciLabware
Azlon®
AZLORPB021PEA 132 EUR
AZLORPB021P AZLORPB031P 212-0085 AZLORPB026P 212-0084 212-0083 212-0082 212-0081 AZLORPB018P
Test tube racks, Azlon®
Gestelle Reagenzglasgestelle
PP
  • Wide range of sizes to fit tubes from 13 to 31 mm diameter
  • Autoclavable and the racks sink in water baths
  • Alpha-numeric place coding
Sturdy test tube racks supplied in self-assembly flat packs.