Printers and Accessories for Mettler Balances
Lieferant: Mettler - Toledo
30094674 30094673
METT30094674EA 1550 EUR
METT30094674 METT30094673
Printers and Accessories for Mettler Balances
Waagen
The BT-P42 printer is a dot matrix printer with Bluetooth® for wireless connection to balance. Requires Bluetooth® interface (BT-multi point or BTS-single point). The P-56RUE and the P-58RUE are thermal printers with real time clock that can simply connect via RS232, USB, Ethernet or Bluetooth with the optional interface. The P-58RUE has more advanced capability including barcode generation, alphanumeric sample ID entry and statistics applications.
A range of accessories is available from Mettler that increases the flexibility and modes of operation. The commonly used items are listedMore Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable