Rampes de minéralisation Kjeldahl
Lieferant: GERHARDT
10-0024 10-0025 10-0026 10-0033 10-0034
705-0473EA 0 EUR
705-0473 458-2349 705-0474 705-0477 705-0478
Rampes de minéralisation Kjeldahl
Aufschlusssysteme Aufschluss-Systeme
Traditional Kjeldahl digestion units for flask sizes from 50 to 750 ml.
  • Adjustable temperature
  • Up to 600 °C for the type KI8/16, power 1800 W
  • Up to 650 °C for the type KI11/26 400 V, 3 phase, power 3480 W
Lieferumfang: Supplied with Kjeldahl flasks, glass exhaust manifold, forked support rods, and power cord but without water jet pump.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable