Lampen, ungefiltert
Lieferant: Vilber Lourmat
312111531 312121531 312131531
VILB312111531EA 0 EUR
VILB312111531 VILB312121531 VILB312131531
Lampen, ungefiltert
Beleuchtung UV-Lampen
Lamps are available from 6 to 40 watt each. They provide a 365 nm UV radiation with visible light.
  • Exclusive UV Master® technology for high UV output
  • Ondulex® reflector for optimum radiation
  • Easy to handle thanks to a table support, handle and wall hook (optional)
* Intensity at one meter
Bestellen
TECHNISCHE DATEN
Environmentally Preferable