Karbolfuchsin, rein
Lieferant: Thermo Scientific
Gefahr
Synonyme: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-yliden)methyl]-o-toluidin Hydrochlorid, 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-yliden)methyl]-o-toluidiniumchlorid, 4-(Di(4-amino-3-methylphenyl)methylen)-2,5-cyclohexadien-1-yliden)ammoniumchlorid, 3,3'-Dimethyl-4,4',4''-triaminotrityliumchlorid
195790050 195790250 195791000
ACRO195790050EA 43.6 EUR
ACRO195790050 ACRO195790250 ACRO195791000
Karbolfuchsin, rein
Karbolfuchsin
Formel: C₂₁H₂₂ClN₃
Molecular Weight: 351,88 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00143923
CAS-Nummer: 4197-24-4
EINECS: 224-086-6
UN: 2923
ADR: 8,III
Bestellen
Spezifikation Testergebnisse
Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
UV Lambda max (in water) about 542 nm
Suitability (for microscopy) Passes test