(+)-Cinchonin ≥98% cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(S)-(+)-[(4S,5R,7R)-5-Ethenyl-1-azabicyclo[2.2.2]octan-7-yl]-quinolin-4-ylmethanol
A17523.14
A17523.22
A17523.14EA
46.1
EUR
A17523.14
A17523.22
(+)-Cinchonin ≥98% cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine
(+)-Cinchonin
Purity: 98+%
CAS No.: 118-10-5
Molecular Formula: C19H22N2O
Formula Weight: 294.39
Melting Point: 253-258°
Beilstein Reference: 89689
Merck Reference: 14,2287
Optical Rotation: +225° (c=0.5 in ethanol)
MDL No.: MFCD00064372
Formel:
C₁₉H₂₂N₂O Molecular Weight: 294,4 g/mol Schmelzpunkt: 253…258 °C Dichte: 1,2 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064372 CAS-Nummer: 118-10-5 EINECS: 204-234-6 Merck Index: 14,2287 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...