Quinine hemisulphate monohydrate 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Quinine sulphate, Quinine sulphate (2:1) (salt) dihydrate
A17036.09
A17036.18
A17036.30
A17036.09EA
43.1
EUR
A17036.09
A17036.18
A17036.30
Quinine hemisulphate monohydrate 99%
Quinine hemisulphate monohydrate
Purity: 99%
CAS No.: 6119-70-6
Molecular Formula: C20H24N2O2.0.5H2SO4.H2O
Formula Weight: 391.47 (373.46anhy)
Odour: Odourless
Melting Point: 217-218° dec.
Beilstein Reference: 6113937
Merck Reference: 14,8061
Optical Rotation: -241° (c=2 in 0.1M HCl)
MDL No.: MFCD00150792
Formel:
C₂₀H₂₄N₂O₂·0.5H₂SO₄·H₂O Molecular Weight: 391,47 g/mol (373,46 anhy) Schmelzpunkt: 217…218 °C (Zers.) Dichte: 1,36 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150792 CAS-Nummer: 6119-70-6 EINECS: 212-359-2 Merck Index: 14,8061 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...