Quinine hemisulphate monohydrate ≥90%, feines weißes Pulver
Lieferant: MP Biomedicals
Achtung
Synonyme: Quinine sulphate, Quinine sulphate (2:1) (salt) dihydrate
0210279250 0210279210 0210279225 0210279280
ICNA0210279250EA 243 EUR
ICNA0210279250 ICNA0210279210 ICNA0210279225 ICNA0210279280
Quinine hemisulphate monohydrate ≥90%, feines weißes Pulver
Quinine hemisulphate monohydrate
Quinine Hemisulfate is an alkaloid originally isolated from the Cinchona family of South American trees. It is an antimalarial agent, anticholinergic, antihypertensive and hypoglycemic agent.
Quinine is an antimalarial agent, anticholinergic, antihypertensive and hypoglycemic agent; also used as a resolving agent. A potassium channel blocker. Also used as a flavoring agent in carbonated beverages, a skeletal muscle relaxant and an antiprotozoal agent for fish. A chiral catalyst used in highly enantioselective [2+2] cycloadditions. A resolving agent for carboxylic acids. Catalyzes the kinetic resolution of furanones.
Potassium channel blocker. Antimalarial, anticholinergic, antihypertensive, and hypoglycemic agent; alkaloid originally isolated from the Cinchona family of South American trees. Inhibits mitochondrial ATP-regulated potassium channel. Used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.
Soluble in water (1 g/810 ml @ 25 °C; 1 g/32 ml @ 100 °C), ethanol (1 g/120 ml @ 25 °C; 1 g/10 ml @ 78 °C); slightly soluble in chloroform or ether. Freely soluble in a mixture of 2 volumes chloroform and 1 volume absolute ethanol.
Formel: C₂₀H₂₄N₂O₂·0.5H₂SO₄·H₂O
Molecular Weight: 391,47 g/mol (373,46 anhy)
Schmelzpunkt: 228 °C (Zers., Lit.)
Dichte: 1,36 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150792
CAS-Nummer: 6119-70-6
EINECS: 212-359-2
Merck Index: 12,03822
Bestellen
Spezifikation Testergebnisse
Identity Test Passes
Purity ≥90%
FTIR Conforms to Standard