2,6-Dichlorphenol-indophenol Natriumsalz Hydrat
Lieferant: Thermo Fisher Scientific
Synonyme: DCPIP Hydrat, Tillmans-Reagenz Hydrat, Dichlorphenolindophenol Hydrat, DPIP Hydrat, Dichlorindophenol Hydrat, DCIP Hydrat, 2,6-Dichlor-N-(4-hydroxyphenyl)-1,4-benzochinonimin Natriumsalz Hydrat, Natrium-4-(3,5-dichlor-4-oxocyclohexa-2,5-dienylidenamino)phenoxid Hydrat
A10107.06 A10107.14 A10107.22
A10107.06EA 58.6 EUR
A10107.06 A10107.14 A10107.22
2,6-Dichlorphenol-indophenol Natriumsalz Hydrat
2,6-Dichlorphenol-indophenol Natriumsalz Hydrat
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase.
It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.
Formel: C₁₂H₆Cl₂NNaO₂·xH₂O
Molecular Weight: 290.08 g/mol (anhy)
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150014
CAS-Nummer: 1266615-56-8
EINECS: 210-640-4