2,6-Dichlorphenol-indophenol Natriumsalz Hydrat 98+%
Lieferant: Thermo Fisher Scientific
Synonyme: DCPIP Hydrat, Tillmans-Reagenz Hydrat, Dichlorphenolindophenol Hydrat, DPIP Hydrat, Dichlorindophenol Hydrat, DCIP Hydrat, 2,6-Dichlor-N-(4-hydroxyphenyl)-1,4-benzochinonimin Natriumsalz Hydrat, Natrium-4-(3,5-dichlor-4-oxocyclohexa-2,5-dienylidenamino)phenoxid Hydrat
152870010 152870100 152870250 152871000
ACRO152870010EA 23.6 EUR
ACRO152870010 ACRO152870100 ACRO152870250 ACRO152871000
2,6-Dichlorphenol-indophenol Natriumsalz Hydrat 98+%
2,6-Dichlorphenol-indophenol Natriumsalz Hydrat
Formel: C₁₂H₆Cl₂NNaO₂·xH₂O
Molecular Weight: 290.08 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150014
CAS-Nummer: 1266615-56-8
EINECS: 210-640-4
Bestellen
Spezifikation Testergebnisse
Appearance Dark green to dark blue-purple Powder
Infrared spectrum Conforms
Loss on drying ≤14 % (1 g, 120°C)
Titration with HClO4 ≥98.0 % (On dry substance)
Suitability (for Ascorbic acid det.) Passes test