ω-Chlor-3,4-dihydroxyacetophenon ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: α-Chlor-3,4-dihydroxyacetophenon, 3,4-Dihydroxyphenacylchlorid, 2-Chlor-3',4'-dihydroxyacetophenon

B23496.14 B23496.22 B23496.36
B23496.14EA 81.7 EUR
B23496.14 B23496.22 B23496.36
ω-Chlor-3,4-dihydroxyacetophenon ≥97%
ω-Chlor-3,4-dihydroxyacetophenon

Purity: 97% CAS No.: 99-40-1 Molecular Formula: C8H7ClO3 Formula Weight: 186.59 Melting Point: 172-176° Beilstein Reference: 2092660

Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
Schmelzpunkt: 172…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002200
CAS-Nummer: 99-40-1
EINECS: 202-754-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR