5'-Chlor-2'-hydroxyacetophenon

Lieferant: CHEMPUR

Synonyme: 1-(5-Chlor-2-hydroxyphenyl)ethan-1-on, 2-Acetyl-4-chlorphenol

FL12069.POR 012069-25G
CHMPFL12069.POREA 0 EUR
CHMPFL12069.POR FLUO012069-25G
5'-Chlor-2'-hydroxyacetophenon
5'-Chlor-2'-hydroxyacetophenon
Formel: C₈H₇ClO₂
Molecular Weight: 170,6 g/mol
MDL: MFCD00067788
CAS-Nummer: 1450-74-4
EINECS: 215-916-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR