5'-Chlor-2'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(5-Chlor-2-hydroxyphenyl)ethan-1-on, 2-Acetyl-4-chlorphenol

M-Clarity™ Qualitätsstufe = MQ100

383384-25G 383384-5G
SIAL383384-25GEA 40.8 EUR
SIAL383384-25G SIAL383384-5G
5'-Chlor-2'-hydroxyacetophenon, Sigma-Aldrich®
5'-Chlor-2'-hydroxyacetophenon
Formel: C₈H₇ClO₂
MDL: MFCD00067788
CAS-Nummer: 1450-74-4
EINECS: 215-916-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR