(±)-Camphen 75% remainder mainly alpha-fenchene

Lieferant: Thermo Scientific
Achtung

Synonyme: Camphen, (±)-2,2-Dimethyl-3-methylenbicyclo[2.2.1]heptan, (±)-3,3-Dimethyl-2-methylennorcamphan

295570025 295570010
ACRO295570025EA 197 EUR
ACRO295570025 ACRO295570010
(±)-Camphen 75% remainder mainly alpha-fenchene
(±)-Camphen
Formel: C₁₀H₁₆
Molecular Weight: 136,24 g/mol
Siedepunkt: 159…160 °C (760 mmHg)
Schmelzpunkt: 42 °C
Dichte: 0,8421 g/cm³ (20 °C)
Flammpunkt: 36 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066603
CAS-Nummer: 79-92-5
EINECS: 201-234-8
UN: 1325
ADR: 4.1,II
Merck Index: 12,01777

Bestellen

Spezifikation Testergebnisse

Appearance White Low melting crystalline solid
Infrared spectrum Conforms
GC ≥70 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR