Azomethin H Hydrat

Lieferant: Thermo Scientific

Synonyme: 8-Hydroxy-1-(salicylidenamino)naphthalen-3,6-disulfonsäure Hydrat, 4-Hydroxy-5-(salicylidenamino)naphthalin-2,7-disulfonsäure Hydrat, Azomethine-H Hydrat, 4-Hydroxy-5-[[(2-hydroxyphenyl)methylen]amino]naphthalin-2,7-disulfonsäure Hydrat, 8-Hydroxy-1-[[(2-hydroxyphenyl)methylen]amino]naphthalin-3,6-disulfonsäure Hydrat

16810.04 16810.09 16810.14
16810.04EA 72.1 EUR
16810.04 16810.09 16810.14
Azomethin H Hydrat
Azomethin H Hydrat

CAS No.: 32266-60-7 Molecular Formula: C17H13NO8S2.xH2O Formula Weight: 423.40(anhy) Physical Form: Crystalline MDL No.: MFCD00066536

Formel: C₁₇H₁₃NO₈S₂·H₂O
Molecular Weight: 441,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066536
CAS-Nummer: 32266-60-7
EINECS: 250-975-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR