Resazurin Natriumsalz
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
7-Natriooxy-3H-phenoxazin-3-on-10-oxid
B21187.03
B21187.06
B21187.03EA
34.1
EUR
B21187.03
B21187.06
Resazurin Natriumsalz
Resazurin Natriumsalz
CAS No.: 62758-13-8
Molecular Formula: C12H6NNaO4
Formula Weight: 251.18
Physical Form: Powder
Odour: Odourless
Beilstein Reference: 3794971
Merck Reference: 14,8141
MDL No.: MFCD00005036
As indicator pH 3.8 (orange) to 6.5 (dark violet)
Formel:
C₁₂H₆NNaO₄ Molecular Weight: 251,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005036 CAS-Nummer: 62758-13-8 EINECS: 263-718-5 Merck Index: 14,8141 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...