9-Fluorenon-4-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 9-Oxofluoren-4-carbonsäure

M-Clarity™ Qualitätsstufe = MQ100

F1557-25G F1557-5G
SIALF1557-25GEA 430 EUR
SIALF1557-25G SIALF1557-5G
9-Fluorenon-4-carbonsäure, Sigma-Aldrich®
9-Fluorenon-4-carbonsäure
Formel: C₁₄H₈O₃
Molecular Weight: 224,22 g/mol
MDL: MFCD00001145
CAS-Nummer: 6223-83-2
EINECS: 228-311-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR