Kupfer(II)phthalocyanin

Lieferant: Thermo Scientific

Synonyme: [29H,31H-Phthalocyaninato-N29,N30,N31,N32]Kupfer, CuPc, Phthalocyanin-Kupfer(II)-Komplex, Phthalocyaninblau, Tetrabenzo-5,10,15,20-diazaporphyrinphthalocyanin, Pigment Blue 15, Kupfer-Phthalocyanin, Heliogenblau B

43650.09 43650.30 43650.18
43650.09EA 60.3 EUR
43650.09 43650.30 43650.18
Kupfer(II)phthalocyanin
Copper(II) phthalocyanine

CAS No.: 147-14-8 Molecular Formula: C32H16CuN8 Formula Weight: 576.08 Physical Form: Powder Appearance: Blue to purple Melting Point: 350° Beilstein Reference: 4121848 Merck Reference: 14,2520 MDL No.: MFCD00010719

Formel: C₃₂H₁₆CuN₈
Molecular Weight: 576,08 g/mol
Schmelzpunkt: 350 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010719
CAS-Nummer: 147-14-8
EINECS: 205-685-1
Merck Index: 14,2520

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR