di-Natrium-α-glycerophosphat Hydrat Gemisch aus Isomeren, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
ortho-Phosphorsäure(2,3-dihydroxypropyl)ester Dinatriumsalz Hydrat
M-Clarity™ Qualitätsstufe = MQ100
G6501-100G
G6501-1KG
G6501-25G
G6501-500G
SIALG6501-100GEA
132
EUR
SIALG6501-100G
SIALG6501-1KG
SIALG6501-25G
SIALG6501-500G
di-Natrium-α-glycerophosphat Hydrat Gemisch aus Isomeren, Sigma-Aldrich®
di-Natrium-α-glycerophosphat Hydrat
Glycerol phosphate disodium salt was used in a novel microplate-based chromatin immunoprecipitation method to immunoprecipitate DNA from rat mesangial cells in a 96-well format.
Usually contains 50% β-isomer and 50% rac-α-isomer
Formel:
C₃H₇Na₂O₆P·H₂O Molecular Weight: 216,04 g/mol (anhy) |
MDL:
MFCD00149082 CAS-Nummer: 55073-41-1 EINECS: 212-464-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...