(1S)-(-)-β-Pinen ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(-)-Pin-2(10)-en
A17818.0F
A17818.AE
A17818.AP
A17818.0FEA
173
EUR
A17818.0F
A17818.AE
A17818.AP
(1S)-(-)-β-Pinen ≥99%
(1S)-(-)-β-Pinen
Purity: 99%
CAS No.: 18172-67-3
Molecular Formula: C10H16
Formula Weight: 136.24
Flash Point: 36°(96°F)
Refractive Index: 1.4780
Odour: Turpentine-like
Melting Point: -61°
Boiling Point: 164-165°
Beilstein Reference: 2038282
Merck Reference: 14,7446
Optical Rotation: -21° (neat)
UN No.: 2319
MDL No.: MFCD00001345
Formel:
C₁₀H₁₆ Molecular Weight: 136.24 g/mol Siedepunkt: 164…165 °C Schmelzpunkt: –61 °C Dichte: 0.860 Flammpunkt: 36 °C (96 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001345 CAS-Nummer: 18172-67-3 EINECS: 242-060-2 UN: 2319 ADR: 3 III Merck Index: 12,07600 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...