2-(Trifluormethyl)benzophenon
Lieferant: FLUOROCHEM
Synonyme:
Phenyl(2-(trifluoromethyl)phenyl)methanone
010686-5G
010686-25G
FLUO010686-5GEA
0
EUR
FLUO010686-5G
FLUO010686-25G
2-(Trifluormethyl)benzophenon
2-(Trifluormethyl)benzophenon
Formel:
C₁₄H₉F₃O Molecular Weight: 250.22 g/mol Siedepunkt: 175…178 °C (31 mmHg) Schmelzpunkt: 62 |
MDL:
MFCD00000377 CAS-Nummer: 727-99-1 EINECS: 211-972-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...