3-(Trifluormethyl)benzophenon

Lieferant: FLUOROCHEM

007594-5G 007594-25G
FLUO007594-5GEA 45.9 EUR
FLUO007594-5G FLUO007594-25G
3-(Trifluormethyl)benzophenon
3-(Trifluormethyl)benzophenon
Formel: C₁₄H₉F₃O
Molecular Weight: 250.22 g/mol
Siedepunkt: 160…162 °C (14 mmHg)
Schmelzpunkt: 54
MDL: MFCD00000389
CAS-Nummer: 728-81-4
EINECS: 211-973-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR