4-(Trifluormethyl)benzophenon

Lieferant: FLUOROCHEM

Synonyme: Phenyl(4-(trifluoromethyl)phenyl)methanone

007595-5G 007595-25G
FLUO007595-5GEA 57.9 EUR
FLUO007595-5G FLUO007595-25G
4-(Trifluormethyl)benzophenon
4-(Trifluormethyl)benzophenon
Formel: C₁₄H₉F₃O
Molecular Weight: 250,22 g/mol
Siedepunkt: 170 °C (21 mmHg)
Schmelzpunkt: 118 °C
MDL: MFCD00000400
CAS-Nummer: 728-86-9
EINECS: 211-974-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR