3-Chlor-2,6-difluorphenol

Lieferant: FLUOROCHEM

005605-5G 005605-1G
FLUO005605-5GEA 0 EUR
FLUO005605-5G FLUO005605-1G
3-Chlor-2,6-difluorphenol
3-Chlor-2,6-difluorphenol
Formel: C₆H₃ClF₂O
Molecular Weight: 164,54 g/mol
MDL: MFCD01631330
CAS-Nummer: 261762-51-0
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR