2-Chlor-3,5-difluorphenol

Lieferant: CHEMPUR

FL9468.1 FL9468.5
CHMPFL9468.1EA 0 EUR
CHMPFL9468.1 CHMPFL9468.5
2-Chlor-3,5-difluorphenol
2-Chlor-3,5-difluorphenol
Formel: C₆H₃ClF₂O
Molecular Weight: 164,54 g/mol
MDL: MFCD00075400
CAS-Nummer: 206986-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR